2-Fluoro-3-methyl-5-pyridineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Fluoro-5-methyl-3-pyridylboronic acid, 2-Fluoro-3-methylpyridine-5-boronic acid, (6-Fluoro-5-methylpyridin-3-yl)boronic acid

BB-8413-5G BB-8413-1G
COBBBB-8413-5GEA 0 GBP
COBBBB-8413-5G COBBBB-8413-1G
2-Fluoro-3-methyl-5-pyridineboronic acid
2-Fluoro-3-methyl-5-pyridineboronic acid
Formula: C₆H₇BFNO₂
MW: 154.94 g/mol
MDL Number: MFCD03411562
CAS Number: 904326-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR