Methyl-3,4-bis(bromomethyl)benzoate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CFF243.1
CFF243.5
017294-1G
017294-2.5G
017294-10G
CHMPCFF243.1EA
0
GBP
CHMPCFF243.1
CHMPCFF243.5
FLUO017294-1G
FLUO017294-2.5G
FLUO017294-10G
Methyl-3,4-bis(bromomethyl)benzoate
Methyl 3,4-bis(bromomethyl)benzoate
Formula:
C₁₀H₁₀Br₂O₂ |
MDL Number:
MFCD00219865 CAS Number: 20896-23-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...