5-Bromo-2-(methylsulphonyl)benzotrifluoride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Bromo-4-(methylsulphonyl)-3-(trifluoromethyl)benzene, 4-Bromo-1-(methylsulphonyl)-2-(trifluoromethyl)benzene
OT-1006-5G
OT-1006-1G
COBBOT-1006-5GEA
0
GBP
COBBOT-1006-5G
COBBOT-1006-1G
5-Bromo-2-(methylsulphonyl)benzotrifluoride
5-Bromo-2-(methylsulphonyl)benzotrifluoride
Formula:
C₈H₆BrF₃O₂S MW: 303.0963 g/mol |
MDL Number:
MFCD13195738 CAS Number: 300356-32-5 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...