Methyl-2-(bromomethyl)-3,5-dinitrobenzoate
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
03-0015.1
03-0015.5
CHMP03-0015.1EA
0
GBP
CHMP03-0015.1
CHMP03-0015.5
Methyl-2-(bromomethyl)-3,5-dinitrobenzoate
Methyl 2-(bromomethyl)-3,5-dinitrobenzoate
Formula:
C₉H₇BrN₂O₆ |
MDL Number:
MFCD04114324 CAS Number: 153754-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...