2-Bromo-4-fluoro-N-methylaniline hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-Methyl 2-bromo-4-fluoroaniline hydrochloride

HC-2876-1G HC-2876-5G
COBBHC-2876-1GEA 0 GBP
COBBHC-2876-1G COBBHC-2876-5G
2-Bromo-4-fluoro-N-methylaniline hydrochloride
2-Bromo-4-fluoro-N-methylaniline hydrochloride
Formula: C₇H₈BrClFN
MW: 240.50 g/mol
MDL Number: MFCD13195687
CAS Number: 1216913-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR