4-(4-Chlorophenoxy)benzaldehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

661309-1G 661309-5G
SIAL661309-1GEA 199 GBP
SIAL661309-1G SIAL661309-5G
4-(4-Chlorophenoxy)benzaldehyde, Sigma-Aldrich®
4-(4-Chlorophenoxy)benzaldehyde
Formula: C₁₃H₉ClO₂
MW: 232.67 g/mol
MDL Number: MFCD01631895
CAS Number: 61343-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR