4'-Cyano-3'-(trifluoromethyl)acetanilide
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CFF186.5
CFF186.10
CFF186.25
004595-25G
004595-5G
CHMPCFF186.5EA
0
GBP
CHMPCFF186.5
CHMPCFF186.10
CHMPCFF186.25
FLUO004595-25G
FLUO004595-5G
4'-Cyano-3'-(trifluoromethyl)acetanilide
4'-Cyano-3'-(trifluoromethyl)acetanilide
Formula:
C₁₀H₇F₃N₂O |
MDL Number:
MFCD00052360 CAS Number: 97760-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...