3,5-Dimethylisoxazole-4-sulphonyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CFF177.5
CFF177.25
017250-5G
017250-25G
017250-1G
CHMPCFF177.5EA
0
GBP
CHMPCFF177.5
CHMPCFF177.25
FLUO017250-5G
FLUO017250-25G
FLUO017250-1G
3,5-Dimethylisoxazole-4-sulphonyl chloride
3,5-Dimethylisoxazole-4-sulphonyl chloride
Formula:
C₅H₆ClNO₃S |
MDL Number:
MFCD00052554 CAS Number: 80466-79-1 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...