Methyl-2-amino-4-(3-chlorophenyl)pyrimidine-5-carboxylate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2272-1G
HC-2272-5G
COBBHC-2272-1GEA
0
GBP
COBBHC-2272-1G
COBBHC-2272-5G
Methyl-2-amino-4-(3-chlorophenyl)pyrimidine-5-carboxylate
Methyl 2-amino-4-(3-chlorophenyl)pyrimidine-5-carboxylate
Formula:
C₁₂H₁₀ClN₃O₂ |
MDL Number:
MFCD12026012 CAS Number: 1150163-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...