2-Amino-4,6-dichloropyrimidine-5-carbaldehyde 96%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde

H27844.03 H27844.06
H27844.03EA 38.3 GBP
H27844.03 H27844.06
2-Amino-4,6-dichloropyrimidine-5-carbaldehyde 96%
2-Amino-4,6-dichloropyrimidine-5-carbaldehyde

Purity: 96% CAS No.: 5604-46-6 Molecular Formula: C5H3Cl2N3O Formula Weight: 192.00 Melting Point: ca 220° MDL No.: MFCD03001242

Formula: C₅H₃Cl₂N₃O
MW: 192.00 g/mol
Melting Pt: ∼ 220 °C
Storage Temperature: Ambient
MDL Number: MFCD03001242
CAS Number: 5604-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR