Ethyl acetate-[1,2-¹³C₂] (99% ¹³C ), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ200

283819-1G 283819-250MG
SIAL283819-1GEA 728 GBP
SIAL283819-1G SIAL283819-250MG
Ethyl acetate-[1,2-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Ethyl acetate-[1,2-13C2]
Formula: C₄H₈O₂
Storage Temperature: Ambient
CAS Number: 84508-45-2
UN: 1173
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR