Accessories for robotic XY module
Supplier: WESBART
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LHT01/12 LHT01/13 LHT01/14 LHT01/16 LHT01/18 LHT01/20 LHT01/24 LHT01/30 LHT01/32 LHT01/36 LHT01/40 LHT01/49 LHT01/60 LHT01/DP10360 LHT01/DP165187
WESBLHT01/12EA 109 GBP
WESBLHT01/12 WESBLHT01/13 WESBLHT01/14 WESBLHT01/16 WESBLHT01/18 WESBLHT01/20 WESBLHT01/24 WESBLHT01/30 WESBLHT01/32 WESBLHT01/36 WESBLHT01/40 WESBLHT01/49 WESBLHT01/60 WESBLHT01/DP10360 WESBLHT01/DP165187
Accessories for robotic XY module
Pumps Peristaltic Pumps
All sectioned container trays and racks for the XY Module are 400×250 mm (l×d) and are made of heavy gauge, anodised aluminium. All corners are welded to contain any spillages.