Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide)

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

011742-100G 011742-25G
FLUO011742-100GEA 0 GBP
FLUO011742-100G FLUO011742-25G
Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide)
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimide)
Formula: C₁₉H₁₅NO₅
MDL Number: MFCD00010733
CAS Number: 82911-69-1
EINECS: 433-520-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR