N-Fmoc-L-leucine

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4003139.0025 4003139.0100 4003139.0500
BACE4003139.0025EA 0 GBP
BACE4003139.0025 BACE4003139.0100 BACE4003139.0500
N-Fmoc-L-leucine
N-Fmoc-L-leucine
Formula: C₂₁H₂₃NO₄
MW: 353.4 g/mol
MDL Number: MFCD00037133
CAS Number: 35661-60-0
EINECS: 252-662-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR