2-Bromo-5-chloro-3-methylbenzo[b]thiophene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

017044-5G 017044-25G 017044-1G
FLUO017044-5GEA 0 GBP
FLUO017044-5G FLUO017044-25G FLUO017044-1G
2-Bromo-5-chloro-3-methylbenzo[b]thiophene
2-Bromo-5-chloro-3-methylbenzo[b]thiophene
Formula: C₉H₆BrClS
MDL Number: MFCD00126363
CAS Number: 175203-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR