Methyl-3-amino-5-fluorobenzo[b]thiophene-2-carboxylate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

022131-25G 022131-5G
FLUO022131-25GEA 0 GBP
FLUO022131-25G FLUO022131-5G
Methyl-3-amino-5-fluorobenzo[b]thiophene-2-carboxylate
Methyl 3-amino-5-fluorobenzo[b]thiophene-2-carboxylate
Formula: C₁₀H₈FNO₂S
MDL Number: MFCD06411643
CAS Number: 835912-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR