Methyl-3-amino-5-nitrobenzo[b]thiophene-2-carboxylate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
011186-1G
011186-5G
FLUO011186-1GEA
0
GBP
FLUO011186-1G
FLUO011186-5G
Methyl-3-amino-5-nitrobenzo[b]thiophene-2-carboxylate
Methyl 3-amino-5-nitrobenzo[b]thiophene-2-carboxylate
Formula:
C₁₀H₈N₂O₄S |
MDL Number:
MFCD00245562 CAS Number: 34674-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...