(Bis(trifluoroacetoxy)iodo)benzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009038-25G 009038-5G
FLUO009038-25GEA 0 GBP
FLUO009038-25G FLUO009038-5G
(Bis(trifluoroacetoxy)iodo)benzene
(Bis(trifluoroacetoxy)iodo)benzene
Formula: C₁₀H₅F₆IO₄
MDL Number: MFCD00009672
CAS Number: 2712-78-9
EINECS: 220-308-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR