4-Aminocyclohexanecarboxylic acid (mixture of cis and trans isomers)
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Aminocyclohexanecarboxylic acid, 4-Aminocyclohexane-1-carboxylic acid
013927-5G
013927-25G
FLUO013927-5GEA
0
GBP
FLUO013927-5G
FLUO013927-25G
4-Aminocyclohexanecarboxylic acid (mixture of cis and trans isomers)
4-Aminocyclohexanecarboxylic acid (mixture of cis and trans isomers)
Formula:
C₇H₁₃NO₂ MW: 143.1800 g/mol |
MDL Number:
MFCD00191601 CAS Number: 1776-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...