Dimethyl cyclohexane-1,4-dicarboxylate (mixture of cis and trans isomers)
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009153-25G
009153-100G
009153-500G
FLUO009153-25GEA
0
GBP
FLUO009153-25G
FLUO009153-100G
FLUO009153-500G
Dimethyl cyclohexane-1,4-dicarboxylate (mixture of cis and trans isomers)
Dimethyl cyclohexane-1,4-dicarboxylate (mixture of cis and trans isomers)
Formula:
C₁₀H₁₆O₄ |
MDL Number:
MFCD00001460 CAS Number: 94-60-0 EINECS: 202-347-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...