Dibenzothiophene-4-boronic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Dibenzothiopheneboronic acid
M-Clarity™ quality level = MQ100
499986-1G
499986-5G
SIAL499986-1GEA
38.8
GBP
SIAL499986-1G
SIAL499986-5G
Dibenzothiophene-4-boronic acid, Sigma-Aldrich®
Dibenzothiophene-4-boronic acid
Formula:
C₁₂H₉BO₂S MW: 228.08 g/mol |
MDL Number:
MFCD01318182 CAS Number: 108847-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...