Benzofuran-2-carboxylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

019325-1G 019325-5G
FLUO019325-1GEA 0 GBP
FLUO019325-1G FLUO019325-5G
Benzofuran-2-carboxylic acid
Benzofuran-2-carboxylic acid
Formula: C₉H₆O₃
MDL Number: MFCD00005848
CAS Number: 496-41-3
EINECS: 207-818-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR