4-Iodo-L-phenylalanine

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: H-Phe(4-I)-OH

4001611.0001 4001611.0005
BACE4001611.0001EA 0 GBP
BACE4001611.0001 BACE4001611.0005
4-Iodo-L-phenylalanine
4-Iodo-L-phenylalanine
Formula: C₉H₁₀INO₂
MW: 291.1 g/mol
MDL Number: MFCD00002602
CAS Number: 24250-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR