3-Bromo-L-phenylalanine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

012748-1G 012748-5G
FLUO012748-1GEA 0 GBP
FLUO012748-1G FLUO012748-5G
3-Bromo-L-phenylalanine
3-Bromo-L-phenylalanine
Formula: C₉H₁₀BrNO₂
MDL Number: MFCD06659110
CAS Number: 82311-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR