4-Fluoro-DL-phenylalanine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002355-100G
002355-25G
002355-5G
FLUO002355-100GEA
0
GBP
FLUO002355-100G
FLUO002355-25G
FLUO002355-5G
4-Fluoro-DL-phenylalanine
4-Fluoro-DL-phenylalanine
Formula:
C₉H₁₀FNO₂ MW: 183.18 g/mol |
MDL Number:
MFCD00002600 CAS Number: 51-65-0 EINECS: 200-113-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...