3-(3,4-Dihydroxyphenyl)-DL-alanine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
DL-β-3,4-Dihydroxyphenyl-Alanine, DL-DOPA, 3,4-Dihydroxy-DL-phenylalanine
M-Clarity™ quality level = MQ100
D9503-1G
D9503-25G
D9503-5G
SIALD9503-1GEA
31
GBP
SIALD9503-1G
SIALD9503-25G
SIALD9503-5G
3-(3,4-Dihydroxyphenyl)-DL-alanine, Sigma-Aldrich®
3-(3,4-Dihydroxyphenyl)-DL-alanine
Formula:
C₉H₁₁NO₄ |
MDL Number:
MFCD00063060 CAS Number: 63-84-3 EINECS: 200-566-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...