2-(Aminomethyl)-5-methylpyrazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (5-Methylpyrazin-2-yl)methanamine

009009-5G 009009-25G
FLUO009009-5GEA 0 GBP
FLUO009009-5G FLUO009009-25G
2-(Aminomethyl)-5-methylpyrazine
2-(Aminomethyl)-5-methylpyrazine
Formula: C₆H₉N₃
MDL Number: MFCD01314542
CAS Number: 132664-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR