Methyl-3-aminobenzo[b]thiophene-2-carboxylate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 3-aminobenzo[b]thiophene-2-carboxylate

CFF240.1 CFF240.5 017292-1G 017292-5G
CHMPCFF240.1EA 0 GBP
CHMPCFF240.1 CHMPCFF240.5 FLUO017292-1G FLUO017292-5G
Methyl-3-aminobenzo[b]thiophene-2-carboxylate
Methyl 3-aminobenzo[b]thiophene-2-carboxylate
Formula: C₁₀H₉NO₂S
MW: 207.25 g/mol
MDL Number: MFCD00206744
CAS Number: 35212-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR