Sodium laurate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Sodium Laurate, Lauric acid sodium salt, Sodium dodecanoate

M-Clarity™ quality level = MQ200

L9755-100G L9755-5G L9755-25G
SIALL9755-100GEA 683 GBP
SIALL9755-100G SIALL9755-5G SIALL9755-25G
Sodium laurate, Sigma-Aldrich®
Sodium laurate
Formula: C₁₂H₂₃NaO₂
MW: 222.304 g/mol
MDL Number: MFCD00041754
CAS Number: 629-25-4
EINECS: 211-082-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR