Gellan gum, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
G1910-1KG
G1910-250G
G1910-5KG
SIALG1910-1KGEA
815
GBP
SIALG1910-1KG
SIALG1910-250G
SIALG1910-5KG
Gellan gum, Sigma-Aldrich®
Gellan gum
Storage Temperature:
Ambient |
MDL Number:
MFCD00131909 CAS Number: 71010-52-1 EINECS: 275-117-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...