2-(3-Bromophenyl)-5-cyclohexyl-1,3,4-oxadiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6378-5G HC-6378-1G
COBBHC-6378-5GEA 0 GBP
COBBHC-6378-5G COBBHC-6378-1G
2-(3-Bromophenyl)-5-cyclohexyl-1,3,4-oxadiazole
2-(3-Bromophenyl)-5-cyclohexyl-1,3,4-oxadiazole
Formula: C₁₄H₁₅BrN₂O
MDL Number: MFCD09878385
CAS Number: 957065-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR