Diethyl vinylphosphonate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
116130-10G
116130-25G
SIAL116130-10GEA
255
GBP
SIAL116130-10G
SIAL116130-25G
Diethyl vinylphosphonate, Sigma-Aldrich®
Diethyl vinylphosphonate
Formula:
C₆H₁₃O₃P MW: 164.14 g/mol |
MDL Number:
MFCD00009079 CAS Number: 682-30-4 EINECS: 211-663-2 UN: 3278 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...