α-Naphtholphthalein, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,3-Bis(4-hydroxynaphthalen-1-yl)isobenzofuran-1(3H)-one
M-Clarity™ quality level = MQ200
N8257-10G
N8257-1G
N8257-5G
SIALN8257-10GEA
712
GBP
SIALN8257-10G
SIALN8257-1G
SIALN8257-5G
α-Naphtholphthalein, Sigma-Aldrich®
α-Naphtholphthalein
Formula:
C₂₈H₁₈O₄ MW: 418.45 g/mol |
MDL Number:
MFCD00036202 CAS Number: 596-01-0 EINECS: 209-875-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...