(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

42119.02 42119.MC 42119.04
42119.02EA 62.5 GBP
42119.02 42119.MC 42119.04
(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl ≥98%
(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl

Purity: 98% CAS No.: 99646-28-3 Molecular Formula: C48H40P2 Formula Weight: 678.79 Physical Form: Powder Appearance: White Melting Point: 255-257° Optical Rotation: +156°(c=0.5 in benzene) MDL No.: MFCD00269856


Chiral catalyst ligand

Formula: C₄₈H₄₀P₂
Melting Pt: 255…257 °C
Storage Temperature: Ambient
MDL Number: MFCD01311709
CAS Number: 99646-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR