(S)-(+)-6,6'-Dibromo-1,1'-bi-2-naphthol
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CS-9134-5G
CS-9134-1G
COBBCS-9134-5GEA
0
GBP
COBBCS-9134-5G
COBBCS-9134-1G
(S)-(+)-6,6'-Dibromo-1,1'-bi-2-naphthol
(S)-(+)-6,6'-Dibromo-1,1'-bi-2-naphthol
Formula:
C₂₀H₁₂Br₂O₂ |
MDL Number:
MFCD00798290 CAS Number: 80655-81-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...