3-Bromo-2-methox-6-methylypyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Bromo-2-methoxy-6-methylpyridine

PY-1387-5G PY-1387-1G
COBBPY-1387-5GEA 0 GBP
COBBPY-1387-5G COBBPY-1387-1G
3-Bromo-2-methox-6-methylypyridine
3-Bromo-2-methox-6-methylypyridine
Formula: C₇H₈BrNO
MW: 202.0485 g/mol
MDL Number: MFCD04112515
CAS Number: 717843-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR