3-Bromo-2-methox-5-(trifluoromethyl)ypyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Bromo-2-methox-5-(trifluoromethyl)ypyridine, 3-Bromo-2-methoxy-5-(trifluoromethyl)pyridine

PY-7341-5G PY-7341-25G PY-7341-1G
COBBPY-7341-5GEA 0 GBP
COBBPY-7341-5G COBBPY-7341-25G COBBPY-7341-1G
3-Bromo-2-methox-5-(trifluoromethyl)ypyridine
3-Bromo-2-methox-5-(trifluoromethyl)ypyridine
Formula: C₇H₅BrF₃NO
MW: 256.02 g/mol
MDL Number: MFCD09972202
CAS Number: 124432-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR