1-(2-Naphthoyl)-3,3,3-trifluoroacetone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003512-5G 003512-25G 003512-100G
FLUO003512-5GEA 0 GBP
FLUO003512-5G FLUO003512-25G FLUO003512-100G
1-(2-Naphthoyl)-3,3,3-trifluoroacetone
1-(2-Naphthoyl)-3,3,3-trifluoroacetone
Formula: C₁₄H₉F₃O₂
MDL Number: MFCD00054501
CAS Number: 893-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR