Ammonium 8-anilinonaphthalene-1-sulphonate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ammonium 8-(phenylamino)naphthalene-1-sulphonate, 8-Anilino-1-naphthalenesulphonic acid Ammonium salt, Ammonium 8-Anilino-1-naphthalenesulphonate, 8-Anilinonaphthalene-1-sulphonic acid ammonium salt, ANS-NH4

10417-100G-F 10417-25G-F 10417-5G-F
SIAL10417-100G-FEA 770 GBP
SIAL10417-100G-F SIAL10417-25G-F SIAL10417-5G-F
Ammonium 8-anilinonaphthalene-1-sulphonate, Sigma-Aldrich®
Ammonium 8-anilinonaphthalene-1-sulphonate
Formula: C₁₆H₁₆N₂O₃S
MDL Number: MFCD00012560
CAS Number: 28836-03-5
EINECS: 249-265-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR