4-Nitrophenyl phosphorodichloridate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
155403-25G
155403-5G
SIAL155403-25GEA
279
GBP
SIAL155403-25G
SIAL155403-5G
4-Nitrophenyl phosphorodichloridate, Sigma-Aldrich®
4-Nitrophenyl phosphorodichloridate
Formula:
C₆H₄Cl₂NO₄P |
CAS Number:
777-52-6 EINECS: 212-289-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...