4-Bromo-8-methyl-2-(trifluoromethyl)quinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2766-5G HC-2766-1G
COBBHC-2766-5GEA 0 GBP
COBBHC-2766-5G COBBHC-2766-1G
4-Bromo-8-methyl-2-(trifluoromethyl)quinoline
4-Bromo-8-methyl-2-(trifluoromethyl)quinoline
Formula: C₁₁H₇BrF₃N
MDL Number: MFCD12026047
CAS Number: 260973-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR