4-Bromo-6-methyl-2-(trifluoromethyl)quinoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2223-5G
HC-2223-1G
COBBHC-2223-5GEA
0
GBP
COBBHC-2223-5G
COBBHC-2223-1G
4-Bromo-6-methyl-2-(trifluoromethyl)quinoline
4-Bromo-6-methyl-2-(trifluoromethyl)quinoline
Formula:
C₁₁H₇BrF₃N |
MDL Number:
MFCD11855862 CAS Number: 18706-27-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...