6-Bromo-8-(trifluoromethyl)quinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2053-5G HC-2053-1G
COBBHC-2053-5GEA 0 GBP
COBBHC-2053-5G COBBHC-2053-1G
6-Bromo-8-(trifluoromethyl)quinoline
6-Bromo-8-(trifluoromethyl)quinoline
Formula: C₁₀H₅BrF₃N
MDL Number: MFCD11504884
CAS Number: 1065074-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR