4-Chloro-8-fluoroquinoline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF953.1 005482-1G 005482-500MG
CHMPCFF953.1EA 0 GBP
CHMPCFF953.1 FLUO005482-1G FLUO005482-500MG
4-Chloro-8-fluoroquinoline
4-Chloro-8-fluoroquinoline
Formula: C₉H₅ClFN
MDL Number: MFCD00278785
CAS Number: 63010-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR