8-Bromo-6-fluoroquinoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7348-1G
PY-7348-5G
COBBPY-7348-1GEA
0
GBP
COBBPY-7348-1G
COBBPY-7348-5G
8-Bromo-6-fluoroquinoline
8-Bromo-6-fluoroquinoline
Formula:
C₉H₅BrFN |
MDL Number:
MFCD09907837 CAS Number: 22960-18-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...