6-Bromo-8-fluoroquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6978-5G HC-6978-1G
COBBHC-6978-5GEA 0 GBP
COBBHC-6978-5G COBBHC-6978-1G
6-Bromo-8-fluoroquinoline
6-Bromo-8-fluoroquinoline
Formula: C₉H₅BrFN
MW: 226.0500 g/mol
MDL Number: MFCD11053791
CAS Number: 220513-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR