2-Bromo-5-(trifluoromethyl)benzyl alcohol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Bromo-5-(trifluoromethyl)benzyl Alcohol
013791-1G
013791-5G
013791-25G
FLUO013791-1GEA
0
GBP
FLUO013791-1G
FLUO013791-5G
FLUO013791-25G
2-Bromo-5-(trifluoromethyl)benzyl alcohol
2-Bromo-5-(trifluoromethyl)benzyl alcohol
Formula:
C₈H₆BrF₃O |
MDL Number:
MFCD04973761 CAS Number: 869725-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...