4-Benzyloxy-3-bromobenzotrifluoride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-1102-5G
OT-1102-25G
OT-1102-1G
COBBOT-1102-5GEA
0
GBP
COBBOT-1102-5G
COBBOT-1102-25G
COBBOT-1102-1G
4-Benzyloxy-3-bromobenzotrifluoride
4-Benzyloxy-3-bromobenzotrifluoride
Formula:
C₁₄H₁₀BrF₃O |
MDL Number:
MFCD11856016 CAS Number: 200956-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...