4-Acetamidobenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018144-1G 018144-5G
FLUO018144-1GEA 0 GBP
FLUO018144-1G FLUO018144-5G
4-Acetamidobenzyl alcohol
4-Acetamidobenzyl alcohol
Formula: C₉H₁₁NO₂
MDL Number: MFCD00016868
CAS Number: 16375-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR