3-Chloro-4,5-diaminobenzotrifluoride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Chloro-5-trifluoromethyl-o-phenylenediamine
004385-1G
004385-5G
FLUO004385-1GEA
0
GBP
FLUO004385-1G
FLUO004385-5G
3-Chloro-4,5-diaminobenzotrifluoride
3-Chloro-4,5-diaminobenzotrifluoride
Formula:
C₇H₆ClF₃N₂ |
MDL Number:
MFCD00042198 CAS Number: 132915-80-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...